CoorsTek™ High-Form Porcelain Crucibles
Withstand temperatures up to 1150°C (2102°F), not to exceed 200°C/hour
Supplier: CoorsTek™ CRS66104
Description
- Glazed inside and out except for outside bottom surface and rim
- Covers sold separately
Specifications
Porcelain | |
10 mL |