High-Frequency Tesla Coil
Safe source of high-frequency, high-voltage electricity.
Supplier:  Customer Channels Group (Ccg) 10206
 View more versions of this product
Catalog No. S43505X
Explore available promotions
Includes:
  • Vibrator, two capacitors, cord and plug
  • Primary coil, secondary coil with 400 turns of wire, adjustable spark gap
  • Plastic housing, lamp socket, two radiating antenna plates Neon bulb, discharge electrode, plastic and copper wires
  • Instructions with 19 experiments, operates on 110V
Description
Description
Transmit electricity without wires. Plasma tube for Tesla coil available separately.
  • Arcs to 3 in. (7.6cm)
  • 50,000V potential
  • Fully assembled coil
Specifications
Specifications
220 V
Transmit electricity without wires; Demonstrates fundamental principles of high frequency electrical phenomena
Vibrator, two capacitors, cord and plug, primary coil, secondary coil with 400 turns of wire, Plastic housing, lamp socket, two radiating antenna plates Neon bulb, discharge electrode, Plastic and Copper wires, instructions with 19 experiments
Tesla Coil
Videos
SDS
Documents
Documents
Accessories