Fisherbrand™ High Precision Metal Scalpels
Fisherbrand Standard Metal Scalpel is nickel-plated for long lasting use.
$18.41 - $18.41
Specifications
Blade Material | Metal |
---|---|
Product Type | Dissecting Scalpel |
For Use With (Application) | For Highly Polished Easy to Clean |
Sterility | Sterile |
Catalog Number | Mfr. No. | Size | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity | ||||||
08-920B
|
N/A | 3.8 cm |
Each for $18.41
Case of 12 Each for $220.92
|
|
||||||
Description
- Fisherbrand™ Standard Metal Scalpel is nickel-plated for long lasting use.
- Nickel-plated construction - constructed to last long
- Easy to clean and sterilize
- Reusable
- Smooth, antiglare finish handle
- Useful in educational, research or healthcare setting
- Fully Autoclavable
- Latex-free
Specifications
Metal | |
Dissecting Scalpel | |
For Highly Polished Easy to Clean | |
Sterile |