Thermo Scientific™ High Temperature Ferrule Replacement Pk. for Exactive GC-Orbitrap GC-MS System
Allows for leak-free, high temperature transferline operation on Orbitrap-GC/MS with the ease-of-use of Vespel ferrules.
Supplier:  Thermo Scientific™ 290VT221
Catalog No. 03-251-523
Explore available promotions
Includes: Replacement Ferrules only
Description
Description
Use the Thermo Scientific™ GC-Orbitrap™ High Temperature Nut and Ferrule Kit to obtain leak-free results at high temperature transferline settings on your gas chromatography (GC) Orbitrap. The high-temperature SCP-5000 Vespel ferrules (included) allow continuous operating transferline temperatures of at least 320°C without ferrule deformation or breakdown (common with standard graphite/Vespel ferrules above 280°C), preventing leaks from forming over an extended period of time. The included spring loaded nut ensures a close fit between nut, ferrule and transferline, further preventing leaks from forming.
High-temperature SCP-5000 Vespel ferrules with 0.4mm ID can be used with most 0.1–0.25mm ID gas chromatography columns. Ferrules must be used in conjunction with High Temperature Adjustable Transferline Nut for GC-Orbitrap (included in kit). Nut and ferrules are for use with the Thermo Scientific™ Q Exactive™ GC Orbitrap™ GC-MS/MS system and the Thermo Scientific™ Exactive™ GC Orbitrap™ GC-MS system.
Specifications
Specifications
Thermo Scientific Q Exactive GC Orbitrap GC-MS/MS System and Thermo Scientific Exactive GC Orbitrap GC-MS System
Vespel
High Temperature Ferrule Replacement Pk.
0.4 mm (Inner), 3.2 mm (Outer)
High Temperature Ferrule Replacement Pk.
2.7 mm
Pack of 5