missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Cytiva His Buffer Kit

Purify histidine-tagged proteins under native conditions. His Buffer Kit consists of convenient, premade buffer stock solutions optimized for histidine-tagged protein purification.

Supplier:  Cytiva 11003400

Catalog No. 45-000-007


Explore available promotions

Includes: 8X phosphate stock solution (pH 7.4), 2 x 100mL; 2M imidizole ( pH 7.4), 100mL

Description

Description

  • High-quality buffers with minimal absorbance at 280nm
  • Optimized protocols for His GraviTrap, His SpinTrap, His MultiTrap HP, His MultiTrap FF and HisTrap columns

Specifications

Specifications

His Buffer Kit
4°C to 30°C
2 x 100mL Phosphate 8x stock solution (pH 7.4), 100mL highly pure 2 M Imidazole (pH 7.4), instructions for use
Each
Videos
SDS
Documents

Documents

Promotions

Promotions