Cytiva HiScreen™ Capto™ SP ImpRes Prepacked IEX Columns
Ideal for method optimization and parameter screening
Supplier: Cytiva 17546815
Description
- Prepacked with Capto SP ImpRes, a strong cation exchanger
- High-flow agarose matrix
- Small bed volume of 4.7mL gives fast results and minimal sample/buffer consumption
- Easily connected in series to achieve 20cm bed height
- Bed height of 10cm to allow method optimization and parameter screening
Specifications
Capto SP ImpRes |