missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Histamine dihydrochloride, 99%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 150620250

Catalog No. AC150620250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Histamine dihydrochloride
0.5% (105°C) max.
0.99
C5H11Cl2N3
25g
154756
Solubility in water: freely soluble in water. Other solubilities: soluble in ethanol,freely soluble in methanol
[H+].[H+].[Cl-].[Cl-].NCCC1=CN=CN1
184.06
184.07
56-92-8
Authentic
Glass bottle
MFCD00012703
25315
histamine dihydrochloride, histamine 2hcl, 2-1h-imidazol-4-yl ethanamine dihydrochloride, 1h-imidazole-4-ethanamine dihydrochloride, 1h-imidazole-4-ethanamine, dihydrochloride, peremin, ceplene, 2-1h-imidazol-4-yl ethylamine dihydrochloride, 2-4-imidazolyl ethylamine dihydrochloride, histaminedium dichloride
PPZMYIBUHIPZOS-UHFFFAOYSA-N
2-(1H-imidazol-5-yl)ethanamine;dihydrochloride
5818
99%
Videos
Safety and Handling

Safety and Handling

Causes serious eye irritation, May cause respiratory irritation, May cause allergy or asthma symptoms or breathing difficulties if inhaled, May cause an allergic skin reaction, Causes skin irritation

Serious eye damage/eye irritation (category 2), Specific target organ toxicity after single exposure (category 3), Respiratory sensitisation (category 1), Skin sensitisation (category 1), Skin corrosion/irritation (category 2)

EINECSNumber : 200-298-4

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only