missing translation for 'onlineSavingsMsg'
Learn More

Histamine dihydrochloride, 99%

CAS: 56-92-8 | C5H11Cl2N3 | 184.06 g/mol

$123.18 - $123.18

Chemical Identifiers

CAS 56-92-8
Molecular Formula C5H11Cl2N3
Molecular Weight (g/mol) 184.06
MDL Number MFCD00012703
InChI Key PPZMYIBUHIPZOS-UHFFFAOYSA-N
Synonym histamine dihydrochloride, histamine 2hcl, 2-1h-imidazol-4-yl ethanamine dihydrochloride, 1h-imidazole-4-ethanamine dihydrochloride, 1h-imidazole-4-ethanamine, dihydrochloride, peremin, ceplene, 2-1h-imidazol-4-yl ethylamine dihydrochloride, 2-4-imidazolyl ethylamine dihydrochloride, histaminedium dichloride
PubChem CID 5818
IUPAC Name 2-(1H-imidazol-5-yl)ethanamine;dihydrochloride
SMILES [H+].[H+].[Cl-].[Cl-].NCCC1=CN=CN1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC150620050
SDS View Documents
Thermo Scientific Chemicals
150620050
5 g Glass bottle
Each for $123.18
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

56-92-8
184.06
PPZMYIBUHIPZOS-UHFFFAOYSA-N
5818
[H+].[H+].[Cl-].[Cl-].NCCC1=CN=CN1
C5H11Cl2N3
MFCD00012703
histamine dihydrochloride, histamine 2hcl, 2-1h-imidazol-4-yl ethanamine dihydrochloride, 1h-imidazole-4-ethanamine dihydrochloride, 1h-imidazole-4-ethanamine, dihydrochloride, peremin, ceplene, 2-1h-imidazol-4-yl ethylamine dihydrochloride, 2-4-imidazolyl ethylamine dihydrochloride, histaminedium dichloride
2-(1H-imidazol-5-yl)ethanamine;dihydrochloride

Specifications

56-92-8
White
Authentic
Glass bottle
MFCD00012703
25315
histamine dihydrochloride, histamine 2hcl, 2-1h-imidazol-4-yl ethanamine dihydrochloride, 1h-imidazole-4-ethanamine dihydrochloride, 1h-imidazole-4-ethanamine, dihydrochloride, peremin, ceplene, 2-1h-imidazol-4-yl ethylamine dihydrochloride, 2-4-imidazolyl ethylamine dihydrochloride, histaminedium dichloride
PPZMYIBUHIPZOS-UHFFFAOYSA-N
2-(1H-imidazol-5-yl)ethanamine;dihydrochloride
5818
99%
Histamine dihydrochloride
249.0°C to 252.0°C
0.5% (105°C) max.
0.99
C5H11Cl2N3
5 g
154756
Solubility in water: freely soluble in water. Other solubilities: soluble in ethanol,freely soluble in methanol
[H+].[H+].[Cl-].[Cl-].NCCC1=CN=CN1
184.06
184.07
Crystalline Powder
Videos
Safety and Handling

Safety and Handling

Causes serious eye irritation, May cause respiratory irritation, May cause allergy or asthma symptoms or breathing difficulties if inhaled, May cause an allergic skin reaction, Causes skin irritation

Serious eye damage/eye irritation (category 2), Specific target organ toxicity after single exposure (category 3), Respiratory sensitisation (category 1), Skin sensitisation (category 1), Skin corrosion/irritation (category 2)

EINECSNumber : 200-298-4

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only