Learn More
Azer Scientific Histology Tissue Control Slides
Designed for use in monitoring stain performance in the pathology laboratory
$270.60 - $727.97
Specifications
| For Use With (Application) | Designed for use in monitoring stain performance in the pathology laboratory |
|---|---|
| Includes | 10 unstained and 1 stained unifrost slide |
| Quantity | 11 Pack |
| Catalog Number | Mfr. No. | Type | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Type | Price | Quantity | |||||
|
22-026-731
|
Azer Scientific
ESCS178 |
Ki67 |
Pack of 10 Each for $592.10
|
|
|||||
|
22-026-718
|
Azer Scientific
ESCS122 |
Melanin Standard |
Pack of 11 Each for $270.60
|
|
|||||
|
22-026-725
|
Azer Scientific
ESCS170 |
PIN-4 |
Pack of 10 Each for $727.97
|
|
|||||
|
22-026-722
|
Azer Scientific
ESCS127 |
Reticulum Standard |
Pack of 10 Each for $312.33
|
|
|||||
|
22-026-702
|
Azer Scientific
ESCS103 |
Pneumocystis Standard |
Pack of 11 Each for $312.33
|
|
|||||
|
22-026-715
|
Azer Scientific
ESCS118 |
Iron Standard Histology Control Slide |
Pack of 11 Each for $278.73
|
|
|||||
|
22-026-713
|
Azer Scientific
ESCS116 |
Glycogen Standard |
Pack of 10 Each for $303.23
|
|
|||||
|
22-026-732
|
Azer Scientific
ESCS179 |
Normal Cervix |
Pack of 10 Each for $574.83
|
|
|||||
|
22-026-704
|
Azer Scientific
ESCS106 |
Amyloid Standard |
Pack of 11 Each for $484.03
|
|
|||||
|
22-026-706
|
Azer Scientific
ESCS109 |
Bile Standard Histology Control Slide |
Pack of 11 Each for $469.93
|
|
|||||
|
22-026-724
|
Azer Scientific
ESCS130 |
Helicobacter |
Pack of 10 Each for $574.83
|
|
|||||
|
22-026-705
|
Azer Scientific
ESCS107 |
Argentaffin Standard |
Pack of 10 Each for $383.23
|
|
|||||
|
22-026-728
|
Azer Scientific
ESCS175 |
ER Immunohistochemistry Control Slide |
Pack of 10 Each for $574.83
|
|
|||||
|
22-026-727
|
Azer Scientific
ESCS174 |
PSA |
Pack of 10 Each for $574.83
|
|
|||||
|
22-026-721
|
Azer Scientific
ESCS126 |
PTAH Standard Histology Control Slide |
Pack of 10 Each for $502.53
|
|
|||||
|
22-026-729
|
Azer Scientific
ESCS176 |
PR Immunohistochemistry Control Slide |
Pack of 10 Each for $574.83
|
|
|||||
|
22-026-719
|
Azer Scientific
ESCS124 |
Mucicarmine Standard |
Pack of 10 Each for $270.60
|
|
|||||
|
22-026-717
|
Azer Scientific
ESCS121 |
Massons Standard |
Pack of 10 Each for $270.60
|
|
|||||
|
22-026-733
|
Azer Scientific
ESCS180 |
HPV |
Pack of 10 Each for $574.83
|
|
|||||
|
22-026-723
|
Azer Scientific
ESCS129 |
Trichrome Standard |
Pack of 10 Each for $312.33
|
|
|||||
|
22-026-720
|
Azer Scientific
ESCS125 |
Periodic Acid Standard |
Pack of 10 Each for $270.60
|
|
|||||
|
22-026-703
|
Azer Scientific
ESCS104 |
Alcian Blue Standard |
Pack of 10 Each for $394.73
|
|
|||||
|
22-026-716
|
Azer Scientific
ESCS120 |
Luxol Fast Blue Standard |
Pack of 10 Each for $303.23
|
|
|||||
|
22-026-711
|
Azer Scientific
ESCS114 |
Fungi-GMS Standard |
Pack of 10 Each for $278.73
|
|
|||||
|
22-026-712
|
Azer Scientific
ESCS115 |
Giemsa Standard |
Pack of 10 Each for $312.33
|
|
|||||
|
22-026-708
|
Azer Scientific
ESCS111 |
Elastic Tissue Standard |
Pack of 10 Each for $270.60
|
|
|||||
Description
- Prepared with paraffin-embedded human tissue sections
- For QC of laboratory's staining processes
Specifications
| Designed for use in monitoring stain performance in the pathology laboratory | |
| 10 unstained and 1 stained unifrost slide | |
| 11 Pack |