Epredia™ Histoplast Paraffin
Meet the demanding and versatile needs of the laboratory with Epredia™ Histoplast Paraffin.
Supplier: Epredia™ 8332
Description
- Made of paraffin and plastic polymers
- Delivers excellent processing and yields thin, wrinkle-free sections
Packaged in convenient, easy-to-open, resealable 2 lb. bags. Also available in a premeasured paraffin kit for use on the Epredia™ Excelsior™ AS Tissue Processor (waste drawer included).
Specifications
50° to 54°C | |
Paraffin |