Epredia™ HistoScreen™ Cassettes in Tube Packs
Eliminates the need for sponges or bags, ideal for very small biopsy tissue samples. Cassettes are compatible with Epredia PrintMate™ Cassette Printers, have a back-edge lid hinge, and loose-packed lids that are available in 12 color varieties.
$2961.81 - $2961.81
Specifications
Closure Included | Yes |
---|---|
Quantity | 750/Cs. |
Pore Style | Mesh |
Closure Type | Separate Lid, Back Hinge |
PrintMate Compatible | Yes |
Catalog Number | Mfr. No. | Color | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Price | Quantity | |||||
B851001BE
|
Epredia™
B851001BE |
Beige |
Case of 750 Each for $2,961.81
|
|
|||||
B851001BL
|
Epredia™
B851001BL |
Blue |
Case of 750 Each for $2,961.81
|
|
|||||
B851001GN
|
Epredia™
B851001GN |
Green |
Case of 750 Each for $2,961.81
|
|
|||||
B851001GY
|
Epredia™
B851001GY |
Gray |
Case of 750 Each for $2,961.81
|
|
|||||
B851001LA
|
Epredia™
B851001LA |
Lavender |
Case of 750 Each for $2,961.81
|
|
|||||
B851001LG
|
Epredia™
B851001LG |
Light Green |
Case of 750 Each for $2,961.81
|
|
|||||
B851001LI
|
Epredia™
B851001LI |
Lilac |
Case of 750 Each for $2,961.81
|
|
|||||
B851001OR
|
Epredia™
B851001OR |
Orange |
Case of 750 Each for $2,961.81
|
|
|||||
B851001TN
|
Epredia™
B851001TN |
Tan |
Case of 750 Each for $2,961.81
|
|
|||||
B851001WH
|
Epredia™
B851001WH |
White |
Case of 750 Each for $2,961.81
|
|
|||||
B851001YW
|
Epredia™
B851001YW |
Yellow |
Case of 750 Each for $2,961.81
|
|
|||||
Description
Printing
- Surface finish and reinforced print faces are engineered to optimize print quality
- Tube design is precisely engineered to reduce jamming and prevent downtime
Grossing, Processing and Embedding
- Cassette design and flow characteristics tested to ensure efficient reagent exchange
- Opening and closing forces adjusted for ease of use, while safeguarding valuable tissue samples
Sectioning
- Precision engineering for solid connection to microtomes
Archiving
- Cassette surface texture optimizes scannability for archiving
Specifications
Yes | |
Mesh | |
Yes | |
HistoScreen Cassette |
750/Cs. | |
Separate Lid, Back Hinge | |
For Use With PrintMate™ Printers |