Fisherbrand™ Histosette™ II Biopsy Processing/Embedding Cassettes with Combination Lid and Base
Histosette™ II Biopsy Processing/Embedding Cassettes with Combination Lid and Base open and close as often as necessary - cassettes will always relock securely.
$458.34 - $731.93
Specifications
Pore Style Slots
Writing Face Angle 45°
Closure Type Combination Lid
Material Plastic
Product Type Biopsy Cassette
View More Specs
Products 10
Catalog Number Mfr. No. Color Price Quantity  
Catalog Number Mfr. No. Color Price Quantity  
15-182-702J
Accessories View Documents
Fisherbrand™
M49310
Lilac
Case of 1500 Each for $458.34
15-182-702K
Accessories View Documents
Fisherbrand™
M49311FS
Orange
Case of 1500 Each for $497.76
15-182-702L
Accessories View Documents
Fisherbrand™
M49312
Aqua
Case of 1500 Each for $459.34
15-182-702A
Accessories View Documents
Fisherbrand™
M4932FS
White
Case of 1500 Each for $514.26
15-182-702B
Accessories View Documents
Fisherbrand™
M4933FS
Pink
Case of 1500 Each for $458.34
15-182-702C
Accessories View Documents
Fisherbrand™
M4934FS
Green
Case of 1500 Each for $497.76
15-182-702D
Accessories View Documents
Fisherbrand™
M4935FS
Yellow
Case of 1500 Each for $552.32
15-182-702E
Accessories View Documents
Fisherbrand™
M4936FS
Blue
Case of 1500 Each for $534.42
15-182-702F
Accessories View Documents
Fisherbrand™
M4937
Peach
Case of 1500 Each for $459.34
15-182-702H
Accessories View Documents
N/A Gray
Case of 1500 Each for $731.93
Description
Description
  • Disposable, plastic cassettes hold tissue specimens during embedding, processing, and storage
  • Molded from a special high-density polymer which is resistant to the chemical action of histological solvents
  • Efficient flow-through slots maximize fluid exchange and ensure proper drainage
  • One-piece integral lid eliminates the need for separate steel lids
  • The anterior writing surface is slanted at a 45° angle, making the cassette more suitable for use with certain types of labeling instruments
Order Info
Available in 11 noncytotoxic, nonmetallic colors.
Specifications
Specifications
Slots
Combination Lid
Biopsy Cassette
45°
Plastic
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'