Simport™ Scientific Histosette™ II Tissue Processing/Embedding Cassette
Designed to hold tissue specimens during the embedding process, as well as in a storage file. Histosette™ II Tissue Processing/Embedding Cassettes also keep specimens safely submerged in liquid.
Supplier:  Simport™ Scientific M4928
 View more versions of this product
Catalog No. 36-101-9178
Explore available promotions
Description
Description
  • Made of acetal
  • Keeps specimens safely submerged in liquid
  • Resistant to the chemical action of most histological solvents
  • 45° angle anterior writing area
  • One-piece integral lid eliminates need for separate steel lids
  • Back mounted tab allows for an unobstructed anterior marking area
  • Open and close as often as necessary - cassettes will always relock securely
  • Suitable for automated printers
Order Info
Each case contains 3 dispenser boxes of 500 cassettes.
Certifications
CE Marked
Specifications
Specifications
Yes
Tan
Acetal
Tissue Cassette
Videos
SDS
Documents
Documents
Accessories
Promotions
Promotions
For in vitro diagnostic (IVD) use only.