Simport™ Scientific Histosette™ II Tissue Processing/Embedding Cassettes in Quickload™ Stacks (Taped)
Secure specimens during processing with Histosette™ II Tissue Processing/Embedding Cassettes in Quickload™ Stacks (Taped). Keep specimens safely submerged in liquid. The flow-through slots maximize fluid exchange. The one-piece integral lid eliminates the need for separate steel lids.
Supplier: Simport™ Scientific M49210T
Description
- In Quickload™ stacks
- 45° anterior writing area
- Open and close as often as necessary - cassettes will always relock securely
- Back mounted tab allowing an unobstructed anterior marking area
- Suitable for automated printers
- Made of acetal
- Resistant to the chemical action of most histological solvents
Order Info
Each case contains 50 Quickload™ stacks of 40 cassettes.
Certifications
CE-IVD
Specifications
2000/Pk. | |
Lilac | |
Acetal |