Simport™ Scientific Histosette™ II Tissue Processing/Embedding Cassettes in Quickload™ Stacks (Taped)
Secure specimens during processing with Histosette™ II Tissue Processing/Embedding Cassettes in Quickload™ Stacks (Taped). Keep specimens safely submerged in liquid. The flow-through slots maximize fluid exchange. The one-piece integral lid eliminates the need for separate steel lids.
$571.21 - $571.21
Catalog Number | Mfr. No. | Color | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Price | Quantity | |||||
22038048
|
Simport™ Scientific
M4925T |
Yellow |
Case of 2000 Each for $571.21
|
|
|||||
22038051
|
Simport™ Scientific
M4928T |
Tan |
Case of 2000 Each for $571.21
|
|
|||||
22038045
|
Simport™ Scientific
M4922T |
White |
Case of 2000 Each for $571.21
|
|
|||||
22038046
|
Simport™ Scientific
M4923T |
Pink |
Case of 2000 Each for $571.21
|
|
|||||
22038047
|
Simport™ Scientific
M4924T |
Green |
Case of 2000 Each for $571.21
|
|
|||||
22038049
|
Simport™ Scientific
M4926T |
Blue |
Case of 2000 Each for $571.21
|
|
|||||
22038050
|
Simport™ Scientific
M4927T |
Peach |
Case of 2000 Each for $571.21
|
|
|||||
22038052
|
Simport™ Scientific
M4929T |
Gray |
Case of 2000 Each for $571.21
|
|
|||||
22038053
|
Simport™ Scientific
M49210T |
Lilac |
Case of 2000 Each for $571.21
|
|
|||||
22038054
|
Simport™ Scientific
M49211T |
Orange |
Case of 2000 Each for $571.21
|
|
|||||
22038055
|
Simport™ Scientific
M49212T |
Aqua |
Case of 2000 Each for $571.21
|
|
|||||
Description
- In Quickload™ stacks
- 45° anterior writing area
- Open and close as often as necessary - cassettes will always relock securely
- Back mounted tab allowing an unobstructed anterior marking area
- Suitable for automated printers
- Made of acetal
- Resistant to the chemical action of most histological solvents
Order Info
Each case contains 50 Quickload™ stacks of 40 cassettes.
Certifications
CE-IVD
Specifications
Acetal |