missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cytiva HiTrap™ Chelating HP Columns
Optimize histidine-tagged protein purifications when nickel ions are not the best choice. Cytiva Life Sciences™ HiTrap™ Chelating HP Columns is prepacked with Chelating Sepharose™ High Performance.
$296.99 - $1156.59
Specifications
Buffer | 6M Guanidine Hydrochloride, 8M Urea |
---|---|
Product Type | HiTrap chelating HP column |
Height (English) | 0.98 in. |
Height (Metric) | 25mm |
Ligand Type | Iminodiacetic Acid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | ||||||
45-000-061
|
Cytiva
17040901 |
1 x 5mL |
Each for $296.99
|
|
||||||
45-000-060
|
Cytiva
17040801 |
5 x 1mL |
Pack of 5 Each for $328.29
|
|
||||||
45-000-062
|
Cytiva
17040903 |
5 x 5mL |
Pack of 5 Each for $1,156.59
|
|
||||||
Description
- Uses different kinds of metal ions
- Binding capacity ∽12mg histidine-tagged protein/mL medium
- Metal ion capacity ∽23μmol Cu2+/mL medium
- Prepacked with Chelating Sepharose High Performance media
- Iminoacetic acid coupled to a 6% agarose matrix by ether linkage
Specifications
6M Guanidine Hydrochloride, 8M Urea | |
0.98 in. | |
Iminodiacetic Acid | |
3 to 13 | |
6% Highly Cross-linked Agarose | |
Affinity Column |
HiTrap chelating HP column | |
25mm | |
3 bar | |
34μm | |
4° to 30°C | |
IMAC and subsequent purification of Polyhistidine tagged Proteins |