missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cytiva HiTrap™ Protein L Prepacked Columns
For capture of antibodies and antibody fragments such as Fab fragments, scFv, and Dabs
$371.69 - $1271.39
Specifications
Product Type | HiTrap Protein L column |
---|---|
Height (English) | 0.78 in. |
Height (Metric) | 20mm |
Flow Rate | <500 cm/hr. |
Material | Polypropylene |
Catalog Number | Mfr. No. | Quantity | Diameter (English) Inner | Price | Quantity | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Diameter (English) Inner | Price | Quantity | ||||
45-002-605
|
Cytiva
17547815 |
1 x 5mL | 0.62 in. |
Each for $1,199.44
|
|
||||
45-002-606
|
Cytiva
17547851 |
5 x 1mL | 0.27 in. |
Each for $1,271.39
|
|
||||
45-000-510
|
Cytiva
29048665 |
1 x 1mL | 0.27 in. |
Each for $371.69
|
|
||||
Description
- Prepacked with Capto L affinity chromatography medium (resin)
- Polypropylene column are easily connected in series for increased productivity
- Easy screening and convenient process development
- For use with syringe, peristaltic pump, or chromatography system
- Maximum pressure: 5 bar
- 1 and 5mL columns
BioProcess Media
- Ligand: Recombinant protein L (E. coli), mammalian free
- Matrix: Rigid, highly cross-linked agarose
- Average particle size: 85μm
- pH stability working range: 2 to 10
1mL columns
- Column I.D.: 7mm
- Flow velocity: <500cm/h3
5mL columns
- Column I.D.: 16mm
- Flow velocity: <500cm/h4
Specifications
HiTrap Protein L column | |
20mm | |
Polypropylene | |
5 bar | |
85μm | |
2° to 8°C | |
Affinity chromato graphy medium (Resin) for capture of Antibodies and Antibody fragments such as Fab fragments, scFv and Dabs |
0.78 in. | |
<500 cm/hr. | |
Recombinant Protein L (E. coli), mammalian free | |
2 to 10 | |
Highly Cross-linked Agarose | |
Affinity Column |