missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Thermo Scientific™ HMDS (Hexamethyldisilazane)

Greatly extends the practical range of GC, improving chromatographic results in the silylation of sugars and related substances.

$1283.62 - $1283.62

Specifications

Standard Concentration 98 to 100%
For Use With Deactivating and Coating Chromatographic Supports
Hazards Corrosive, Flammable
Storage Room Temperature
Standard Components Hexamethyldisilazane
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
PI84770
View Documents
Thermo Scientific™
TS84770
25 g
Pack for $1,283.62
 
Description

Description

Thermo Scientific™ HMDS (Hexamethyldisilazane) greatly extends the practical range of GC, improving chromatographic results in the silylation of sugars and related substances.

  • Suitable for deactivating and coating chromatographic supports
  • Monofunctional, making polymerization not possible and eliminating surface moisture

Specifications

Specifications

98 to 100%
Corrosive, Flammable
Hexamethyldisilazane
Deactivating and Coating Chromatographic Supports
Room Temperature
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'

For Research Use Only. Not for use in diagnostic procedures.