Hoefer™ Blotting Paper DFS Item
Cellulose papers precut for convenience
$154.33 - $169.80
Specifications
Format Sheet
Material Cellulose
View More Specs
Products 3
Catalog Number Mfr. No. Length (Metric) Width (Metric) For Use With (Equipment) Price Quantity  
Catalog Number Mfr. No. Length (Metric) Width (Metric) For Use With (Equipment) Price Quantity  
03-500-286 DFS Item
View Documents
Hoefer™
TE26
9 cm 10.5 cm TE22 Mini Tank Blotting Unit or MiniVE Blot Module
Pack of 50 Each for $169.80
03-500-302 DFS Item
View Documents
Hoefer™
TE76
21 cm 25 cm TE77 Semi-Dry Blotting Units
Pack of 25 Each for $166.65
03-500-303 DFS Item
View Documents
Hoefer™
TE761416
14 cm 16 cm TE70 or TE77 Semi-Dry Blotting Units
Pack of 25 Each for $154.33
Description
Description
  • For Western Blotting
Specifications
Specifications
Sheet
Cellulose
Videos
SDS
missing translation for 'documents'
missing translation for 'documents'