missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Hoefer™ SUB13 Midi Submarine Running Tray
Accessories for SUB13 Midi Submarine
Supplier: Hoefer™ SUB13UT
Description
- 12.8 x 15 cm
- Silicone Gasket
Specifications
SUB13 Midi Submarine Gel Electrophoresis Unit | |
Silicone gasket | |
5.9 in. | |
5 in. |
Running Tray | |
5.9 x 5 in. (15 x 12.8 cm) | |
15 cm | |
12.8 cm |