United Scientific™ Hoffman Electrolysis Apparatus DFS Item
Hoffman Electrolysis Apparatus for qualitative and quantitative study of electrolytic decomposition of electrolytes such as water and hydrochloric acid.
Supplier:  United Scientific™ HEA001P
 View more versions of this product
Catalog No. S07010
Explore available promotions
Includes: Support stand, burette clamp, extension clamp, bosshead clamp, pair of platinum electrodes, pair of carbon electrodes, activity guide
Description
Description
  • Tubes are 2 in. (50 mm) each
  • Dimensions: 24 x 12 x 6 in. (61 x 31 x 15 cm)
Specifications
Specifications
Support stand, burette clamp, extension clamp, bosshead clamp, pair of platinum electrodes, pair of Carbon electrodes, activity guide
Replacement Platinum Electrodes
Qualitative and quantitative study of electrolytic decomposition of electrolytes such as Water and hydrochloric Acid
Borosilicate Glass