missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

M+A Matting Hog Heaven™ Anti-Fatigue Mat

Hog Heaven™ Anti-Fatigue Mats are designed to provide comfort and durability in a variety of harsh conditions.

Supplier:  M+A Matting 421000023

 View more versions of this product

Catalog No. 18333134

Explore available promotions

Description

Description

Black
Specifications

Specifications

2 x 3 ft.
Electrically conductive
3 ft.
Rubber borders
2 ft.
Yellow
Chemical-resistant Mat
5/8 in.
Videos
SDS
Documents

Documents

Promotions

Promotions