missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
M+A Matting Hog Heaven™ Anti-Fatigue Mat
Hog Heaven™ Anti-Fatigue Mats are designed to provide comfort and durability in a variety of harsh conditions.
Supplier: M+A Matting 421000312
Description
BlackSpecifications
3 x 12 ft. | |
Electrically conductive | |
12 ft. | |
Rubber borders |
3 ft. | |
Yellow | |
Chemical-resistant Mat | |
5/8 in. |