missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Synthware™ Hollow Penny Head Stopper

Heavy wall hollow penny head stopper

$26.71 - $71.38

Specifications

Description Heavy-wall
Color Clear
Material Borosilicate Glass
For Use With (Application) Organic Synthesis
Quantity 1 Ea.
View More Specs

Products 7
Catalog Number Mfr. No. Price Quantity  
Catalog Number Mfr. No. Price Quantity  
31501869
View Documents
Synthware™
S401410
Each for $26.71
 
31501870
View Documents
Synthware™
S401420
Each for $27.50
 
31501871
View Documents
Synthware™
S401922
Each for $30.21
 
31501872
View Documents
Synthware™
S402440
Each for $31.82
 
31501873
View Documents
Synthware™
S402942
Each for $39.53
 
31501874
View Documents
Synthware™
S403445
Each for $51.12
 
31501875
View Documents
Synthware™
S404550
Each for $71.38
 
Description

Description

Specifications

Specifications

Heavy-wall
Borosilicate Glass
1 Ea.
Clear
Organic Synthesis
SDS
missing translation for 'documents'

missing translation for 'documents'