missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Holmium foil, 0.025mm (0.001in) thick, 99.9% (REO), Thermo Scientific Chemicals

$345.15 - $1205.27

Chemical Identifiers

CAS 7440-60-0
Molecular Formula Ho
Molecular Weight (g/mol) 164.93
MDL Number MFCD00011049
InChI Key KJZYNXUDTRRSPN-UHFFFAOYSA-N
Synonym atom, unii-w1xx32sqn1, w1xx32sqn1, foil, holmio, chips, ingot, pieces, powder, acmc-1c9ym
PubChem CID 23988
ChEBI CHEBI:49648
IUPAC Name holmium
SMILES [Ho]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA42687FF
View Documents
Thermo Scientific Chemicals
042687FF
25 x 25 mm
Each for $345.15
 
AA42687FI
View Documents
Thermo Scientific Chemicals
042687FI
50 x 50 mm
Each for $1,205.27
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged in mylar under argon
Specifications

Chemical Identifiers

7440-60-0
164.93
KJZYNXUDTRRSPN-UHFFFAOYSA-N
23988
holmium
Ho
MFCD00011049
atom, unii-w1xx32sqn1, w1xx32sqn1, foil, holmio, chips, ingot, pieces, powder, acmc-1c9ym
CHEBI:49648
[Ho]

Specifications

Gray to Silver
Ho
UN3178
atom, unii-w1xx32sqn1, w1xx32sqn1, foil, holmio, chips, ingot, pieces, powder, acmc-1c9ym
[Ho]
164.93
CHEBI:49648
Odorless
≈0.14g/25x25mm
Holmium foil, 0.025mm (0.001 in.) thick
7440-60-0
MFCD00011049
25 x 25 mm
KJZYNXUDTRRSPN-UHFFFAOYSA-N
holmium
23988
99.9%
Mylar under Argon
Foil
(REO)
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H228-H260
Flammable solid.
In contact with water releases flammable gases which may ignite spontaneously.

GHS P Statement
P210-P231+P232-P280-P370+P378c-P402+P404-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Handle under inert gas.
Protect from moisture.
Wear protective gloves/protective clothing/eye protection/face protection.
In case of fire: Use for extinction: CO2, sand, extinguishing powder.
Store in a dry place.
Store in a closed container.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLID, INORGANIC, N.O.S.

EINECSNumber : 231-169-0

TSCA : Yes

storageNote1 :

  • Packaged in mylar under argon

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only