Learn More
Homophthalic acid, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 120840250
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Homophthalic acid | |
89-51-0 | |
97.5% min. (HPLC) | |
C9H8O4 | |
MFCD00004326 | |
09, 857 | |
Solubility in water: 12g/L (20 c) | |
C1=CC=C(C(=C1)CC(=O)O)C(=O)O | |
180.16 | |
180.16 |
99% | |
Authentic | |
Glass bottle | |
HO2CCH2C6H4CO2H | |
25g | |
homophthalic acid, 2-carboxymethyl benzoic acid, benzeneacetic acid, 2-carboxy, 2-carboxyphenylacetic acid, 2-carboxybenzeneacetic acid, alpha-carboxy-o-toluic acid, o-carboxymethyl benzoic acid, 2-carboxyphenyl acetic acid, o-toluic acid, alpha-carboxy, toluene-alpha,2-dicarboxylic acid | |
ZHQLTKAVLJKSKR-UHFFFAOYSA-N | |
2-(carboxymethyl)benzoic acid | |
66643 | |
98% |
Chemical Identifiers
89-51-0 | |
180.16 | |
ZHQLTKAVLJKSKR-UHFFFAOYSA-N | |
66643 | |
C1=CC=C(C(=C1)CC(=O)O)C(=O)O |
C9H8O4 | |
MFCD00004326 | |
homophthalic acid, 2-carboxymethyl benzoic acid, benzeneacetic acid, 2-carboxy, 2-carboxyphenylacetic acid, 2-carboxybenzeneacetic acid, alpha-carboxy-o-toluic acid, o-carboxymethyl benzoic acid, 2-carboxyphenyl acetic acid, o-toluic acid, alpha-carboxy, toluene-alpha,2-dicarboxylic acid | |
2-(carboxymethyl)benzoic acid |
Safety and Handling
GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and
GHS Signal Word: Warning
EINECSNumber : 201-913-9
RUO â Research Use Only