missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Homophthalic acid, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 120841000

 View more versions of this product

Catalog No. AC120841000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Homophthalic acid
89-51-0
97.5% min. (HPLC)
C9H8O4
MFCD00004326
09, 857
Solubility in water: 12g/L (20 c)
C1=CC=C(C(=C1)CC(=O)O)C(=O)O
180.16
180.16
99%
Authentic
Glass bottle
HO2CCH2C6H4CO2H
100g
homophthalic acid, 2-carboxymethyl benzoic acid, benzeneacetic acid, 2-carboxy, 2-carboxyphenylacetic acid, 2-carboxybenzeneacetic acid, alpha-carboxy-o-toluic acid, o-carboxymethyl benzoic acid, 2-carboxyphenyl acetic acid, o-toluic acid, alpha-carboxy, toluene-alpha,2-dicarboxylic acid
ZHQLTKAVLJKSKR-UHFFFAOYSA-N
2-(carboxymethyl)benzoic acid
66643
98%

Chemical Identifiers

89-51-0
180.16
ZHQLTKAVLJKSKR-UHFFFAOYSA-N
66643
C1=CC=C(C(=C1)CC(=O)O)C(=O)O
C9H8O4
MFCD00004326
homophthalic acid, 2-carboxymethyl benzoic acid, benzeneacetic acid, 2-carboxy, 2-carboxyphenylacetic acid, 2-carboxybenzeneacetic acid, alpha-carboxy-o-toluic acid, o-carboxymethyl benzoic acid, 2-carboxyphenyl acetic acid, o-toluic acid, alpha-carboxy, toluene-alpha,2-dicarboxylic acid
2-(carboxymethyl)benzoic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and

GHS Signal Word: Warning

EINECSNumber : 201-913-9

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only