missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Honeywell™ Eyesaline™ Personal Eyewash Bottle
A Honewell Eyesaline™ Personal Eyewash Bottle provides immediate relief for eye injuries.
Supplier: Honeywell™ 320004450000
Includes:
Complete instructions for use
Description
Carry-along squeeze bottles of saline solution.
- Ideal for bathing eyes while transporting injured worker to hospital
- Safety sealed and dated
- Single-use only with snap-off tops for quick and easy access
Compliance
Does not meet the criteria of plumbed or self-contained eyewash equipment.
Specifications
8 oz. | |
FDA | |
Buffered, Saline Solution and Superior to Tap Water, Temper Proof Tab | |
Eyewash Bottle |
236.5 mL | |
First Aid Kit and Travel Kit, Agriculture, Chemical, Construction, Food Services, Oil and Gas, Steel, Welding Industries | |
Polyethylene | |
Sterile |