Learn More
Honeywell Safety Products™ Saline Wall Station
The Honeywell Saline Wall Station is easily mounted in hazardous areas to provide immediate relief in case of eye injury.
Supplier: Honeywell Safety Products™ 320004600000H5
Alert:
Since the first few seconds of an eye injury are critical, an eyewash bottle station will provide immediate relief until the worker has access to either a plumbed station or a shower. Eyewash bottles support a plumbed or self-contained drenching facility. They do not take the place of a 15-minute flushing facility.
Description
The Honewell Saline Wall Station is easily mounted in hazardous areas to provide immediate relief in case of eye injury.
Choose from 16 or 32 oz. (473 or 946mL) bottles.
- Highly visible, with full emergency-use instructions on front
- Made from sturdy, high-impact plastic
- Bottles are single-use only with snap-off tops for quick and easy access
- Bottles have extended flow nozzles; 32 oz. bottles have integral grip for ease of use in an emergency
Order Info
Refill bottles sold separately; see 19-003-030.
Compliance
Does not meet the criteria of plumbed or self-contained eyewash equipment.
Specifications
16 oz. | |
FDA | |
For Refill Bottles, See 19-003-030. | |
Polyethylene |
500 mL | |
Flushing Nuisance Particles Such as Pollen, Dust, Saw Dust, Smoke and More | |
Sperian Saline Buffered, Saline Solution Superior to Tap Water | |
Single Eye Wall Station |