missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Honeywell™ WorkEasy™ 300 Dyneema™ Cut-Resistant Gloves, Polyurethane Coated
Dyneema™ fiber blend provides a high level of protection against cuts and abrasions, while still maintaining a bare-handed feel.
$11.76 - $11.76
Specifications
Coating Material | Polyurethane |
---|---|
Color | Gray |
Cut Resistance Rating | ANSI A3 |
Exterior Finish | No Texture |
Hand | Specific |
Catalog Number | Mfr. No. | Size | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity | ||||||
19-175-487
|
Honeywell™
WE300L |
Large |
Pair for $11.76
Case of 144 Pair for $666.72
|
|
||||||
Description
- High level of cut protection for hazardous applications
- Lightweight fabric blend provides the same level of protection, without the bulk to prevent hand fatigue
- Polyurethane coating provides abrasion resistance and enhanced grip
- Launderable for extended glove life
Specifications
Polyurethane | |
ANSI A3 | |
Specific | |
Cut-resistant Gloves | |
White | |
WorkEasy 300 |
Gray | |
No Texture | |
Dyneema Fiber | |
Small parts assembly, fabrication, inspection, warehousing | |
Knit | |
13 |