SP Scientific Hotpack Glassware Washer Accessories
Accessories designed for use with Hotpack™ Undercounter, Freestanding and Vertical Stacking Glassware washers safeguard flasks, beakers, test tubes, bottles, Petri dishes, and more.
Supplier:  SP Scientific 437219
 View more versions of this product
Catalog No. 15365233
Explore available promotions
Description
Description
  • Accessories maximize space, improve efficiency, and protect specialty glassware; use in standard open racks
  • Bottom mesh inserts prevent small items from falling through
  • Made of stainless steel for long life and durability
  • Booster Pump for use with low pressure DI water supply below 18psi
Specifications
Specifications
Stainless Steel
Stainless Steel
Washing Lab Glassware
Fits 1/2 of rack, bottom only
30.5 cm
12 in. x 12 in. x 4 in.
10 cm
30.5 cm
30.5 cm
2.2 kg
Test Tube Basket Insert with Lid
Up to 150, 200mm long test tubes
Lab Glassware Washer
12 in.
12 in. x 12 in.
4 in.
12 in.
12 in.
5 lb.
Each