missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

SP Scientific Hotpack Vertical Stacking Glassware Washers

Designed for ease of operation and flexible loading, Hotpack Vertical SpaceSaver™ Laboratory Glassware Washers provide large load capacity and built-in flexibility, all in a small footprint. Equipped with premium features including a soft touch controller, enhanced drying, and quiet run cycles, SP Hotpack Laboratory Glassware Washers are engineered for durability and energy efficient operation.

Supplier:  SP Scientific VSWHA11001

 View more versions of this product

Catalog No. 15365216

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Explore available promotions

Description

Description

  • Provide reliable day-in, day-out performance so your laboratory glass is always ready-to-go
  • Two independently operated chambers with up to four loading levels for high volume washing
  • All models come with removable top and bottom stainless-steel open racks to accommodate a variety of glassware 
  • Spindle rack ready levels accommodate direct injection spindle racks (sold separately) for thorough cleaning inside narrow-neck glassware
  • Stainless steel baskets, insert racks and accessories sold separately to maximize space, improve efficiency, and protect specialty glassware
  • Temperature selection to 70°C (158°F); can effectively dissolve stubborn oils
  • DI rinse models ensure a spotless finish; easily connect to fixed or portable DI water systems 
  • Optional DI Booster Pump recommended when the incoming DI water source has a pressure of less than 18psi
  • Delayed start feature allows washer to operate unattended during off-peak hours to reduce energy costs
  • Designed for optimal quiet operation with noise buffering insulation
  • Three filters protect pumps from debris; conveniently located for easy access
  • Minimal, efficient water consumption 
  • Enhanced dual fan drying system for spotless results
  • 304 stainless steel chamber and exterior casing ensures corrosion protection and long life
  • 3 in. lockable casters allow easy, controlled mobility of unit 
  • UL 61010-1 and CSA 22.2 61010-1 certified by MET Labs
  • Microprocessor control system features 6 cycle selections including a customizable timed option to ensure minimum energy consumption. All wash cycles have pre-wash, wash, pre-rinse and rinse cycles to prevent contamination. 
  • Rinse Only - Short tap water rinse cycle
  • Normal Wash - Lower temperature (55°C) longer cycle with three (3) final rinse cycles
  • Daily Wash - Standard temperature (65°C) with two (2) final rinse cycles
  • Heavy Wash - Higher temperature (70°C) longer cycle with multiple pre-wash and final rinse cycles
  • Heavy Wash Time Saver - Lower temperature (55°C) version of Heavy Wash with a shorter overall cycle time
  • Timed Wash Programs - Standard temperature (65°C) in programmable 15 minute increments up to 2.5 hours
Specifications

Specifications

DI Rinse - Top and Bottom Standard Racks
Stainless Steel
Per Chamber: Min: Tap 2.4 gal. (9L), DI 0.8 gal. (3L); Max: Tap 4 gal. (15L), DI 1.6 gal. (6L)
Washing Lab Glassware
23.31 in.
78.5 in.
26.38 in.
370 lb.
120V, 60Hz, 15A
60 Hz
Digital
60 psi
MET Labs
55°-70°C (131°-158°F)
59.2 cm
199.4 cm
67 cm
168 kg
120 V Plug in
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions