Justrite™ HPLC Poly Manifold for Quick-Disconnect Disposal Cans
For use with Justrite™ Centura Quick-Disconnect HPLC Safety Disposal Cans
Supplier:  Justrite™ 28179
 View more versions of this product
Catalog No. 19-412-604
Explore available promotions
Description
Description
For use with Justrite™ Centura Quick-Disconnect HPLC Safety Disposal Cans
  • Enables up to six waste lines per manifold from HPLC processes to be fed into a single intake fitting
  • Six 1/4-28 ports and one 3/8 in. NPT port plus caps for unused ports
  • Tapped to accommodate 1/4-28 threads of separately available compression fittings
  • Multiple manifolds may be used on a custom-designed Quick Disconnect container as long as one disconnect remains unused to permit venting
  • Cat. No. 19-412-604 has a “stacker” connection enabling it to attach on top of Cat. No. 19-412-602 or 19-412-603; allows up to 12 lines to be fed into disposal can
  • Fittings sold separately (See 19-412-605, -606)
Specifications
Specifications
Quick-disconnect Disposal Can
For Justrite™ centura quick-disconnect HPLC safety disposal cans
Cap for unused ports, Manifold w/SS stacker connect
7 x 5 x 4.5 in. (17.8 x 12.7 x 11.4 cm)
17.8 cm
12.7 cm
11.4 cm
US OSHA, NFPA Code 30
Stainless Steel, Polypropylene
Ea.
7 in.
5 in.
4.5 in.