Restek™ HPLC Stainless Steel Capillary Tubing Assemblies DFS Item
For Agilent HPLC Systems.
Supplier:  Restek™ RK26537
 View more versions of this product
Catalog No. 06-728-962
Explore available promotions
Description
Description
  • Precut, micropolished tubing and preseated fittings for quick, easy maintenance of your Agilent HPLC systems
  • Material: Stainless steel
Specifications
Specifications
Agilent HPLC Systems
Front and back ferrules
HPLC Systems
0.06 in.
Stainless Steel (316)
Fitting
Smooth
0.0625 in.
0.15 cm