Air-Tite HSW Soft-Ject™ Disposable Syringes
Air-Tite HSW Soft-Ject™ Disposable Syringes are high-quality, three-part disposable rubber piston syringes
Supplier: Air-Tite R20
Description
- Lubricated synthetic rubber piston allows for smooth draw while positive stop helps prevent accidental pullout
- Bold graduations are easily read and resist wipe off
- ETO gas sterilized syringes, latex and pyrogen free
- Packaged in individual sterile strip packs
- Made in Germany
Specifications
Yes | |
Single-use | |
Leur Slip | |
Sterile |
Soft-Ject Syringe | |
20 mL | |
No |