missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Air-Tite HSW Soft-Ject™ Disposable Syringes
Air-Tite HSW Soft-Ject™ Disposable Syringes are high-quality, three-part disposable rubber piston syringes
Supplier: Air-Tite R10
Description
- Lubricated synthetic rubber piston allows for smooth draw while positive stop helps prevent accidental pullout
- Bold graduations are easily read and resist wipe off
- ETO gas sterilized syringes, latex and pyrogen free
- Packaged in individual sterile strip packs
- Made in Germany
Specifications
Yes | |
Single-use | |
10 mL | |
Yes |
Soft-Ject Syringe | |
12 mL | |
Leur Slip | |
Sterile |