Air-Tite™ HSW Soft-Ject™ Disposable Syringes
High-quality, three-part disposable rubber piston syringes
Supplier: Air-Tite™ RL3
Description
High-quality, three-part disposable rubber piston syringes
- Lubricated synthetic rubber piston allows for smooth draw while positive stop helps prevent accidental pullout
- Bold graduations are easily read and resist wipe off
- ETO gas sterilized syringes, latex and pyrogen free
- Packaged in individual sterile strip packs
- Made in Germany
Certifications
FDA registered CE Mark, ISO 9001 certified.
Specifications
FDA, CE, ISO 9001 | |
Soft-Ject™ Syringe | |
3 mL | |
Yes |
Yes | |
Single-use | |
Luer Lock | |
Sterile |