Invitrogen™ Human BCRP Vesicles DFS Item
Human BCRP Vesicles are insect derived purified plasma membranes with inserted BCRP transport protein (Breast Cancer Resistance Protein, ABCG2). Our Human BCRP Vesicles can be used to evaluate test compounds and drug interactions with BCRP in in vitro assays.
Supplier:  Invitrogen™ GM0008
Catalog No. GM0008
Explore available promotions
Description
Description
Specifications
Specifications
Frozen
Research Grade
Human Vesicle
Vial
0.5 mL
Human
Store in ultracold freezer (-60 to -85°C) and protect from light.
Dry Ice
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.