missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Invitrogen™ Human Complement C3a ELISA Kit

ELISA

$816.00 - $2885.00

Specifications

Assay Range 0.3 to 20 ng/mL
Assay Sensitivity 0.14 ng/mL
Interassay CV 5.9%
Intraassay CV 5.9
For Use With (Application) ELISA
View More Specs

Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human C3a

Products 3
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
501125293
View Documents
Invitrogen™
BMS2089
96 Tests
Each for $816.00
 
501125296
View Documents
Invitrogen™
BMS2089TWO
2 x 96 Tests
Each for $1,360.00
 
501125294
View Documents
Invitrogen™
BMS2089FIVE
5 x 96 Tests
Each for $2,885.00
 
Description

Description

The Human Complement C3a (Hu C3a) ELISA quantitates Hu C3a in human serum, plasma or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu C3a. Principle of the method The Human C3a solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

Complement component C3 plays a central role in the activation of complement system. Its activation is required for both classical and alternative complement activation pathways. People with C3 deficiency are susceptible to bacterial infection.
Specifications

Specifications

0.3 to 20 ng/mL
5.9%
ELISA
718
0.14 ng/mL
5.9
Colorimetric Microplate Reader
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'