GIBCO™ Human Cytosol DFS Item
Pooled human liver cytosolic fractions. Total protein concentration: 20mg/mL
Supplier:  GIBCO™ HMCYPL
Catalog No. HMCYPL
Explore available promotions
Description
Description
Specifications
Specifications
Unisex
Pooled
1 mL
Human
Human Cytosol
Adult
Cytosol
Gibco™
Frozen
Liver
Research Grade
20 mg/mL
Store between -60°C and -85°C.
Tube
Dry Ice
Videos
SDS
Documents
Documents
Promotions
Promotions
For Research Use Only. Not for use in diagnostic procedures.