missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Gibco™ Human Endothelial SFM
Human Endothelial-SFM
Supplier: Gibco™ 11111044
Description
Human Endothelial-SFM
Human Endothelial-SFM is a basal serum-free growth medium developed to support the isolation and expansion of human umbilical venous and arterial endothelial cells (1–4). Medium requires supplementation with bFGF (20 ng/mL), EGF (10 ng/mL), and human plasma fibronectin (10μg/mL). Contains L-glutamine.Specifications
Store in refrigerator (2–8°C) and protect from light. | |
Endothelial Cells | |
Mammalian Cell Culture | |
Serum Free Medium (SFM) | |
Sterile-filtered | |
Glutamine, Phenol Red | |
500 mL |
HUAEC, HUVEC | |
Serum-free | |
Liquid | |
Serum-free | |
Sterile-filtered | |
≤10 EU/mL, ≤ 10 EU/mL | |
Room Temperature |
For Research Use Only. Not for use in diagnostic procedures.