Learn More
Gibco™ Human Heat Stable bFGF Recombinant Protein
Recombinant Protein
Supplier: Gibco™ PHG0367
Description
Carrier-Free Reconstitution: Centrifuge the vial briefly, before opening to bring the contents to the bottom. Reconstitute the lyophilized protein in sterile, distilled water to a concentration of 1.0 mg/mL. Alternatively, reconstitute at lower concentrations in carrier-containing medium. Do not vortex. Apportion the reconstituted protein into working aliquots and store at -20°C. Make any further dilutions of the reconstituted protein in medium, or buffered solution containing carrier protein, such as PBS with 0.1% BSA. For the 5 μg unit size, reconstitute directly in carrier-containing medium. For other unit sizes, reconstitute to 1.0 mg/mL as follows: 50 μg, reconstitute in 50 μL 100 μg, reconstitute in 100 μL 500 μg, reconstitute in 500 μL Storage: Store the lyophilized protein at -20°C, preferably desiccated. Upon reconstitution, apportion into working aliquots and store at -20°C (not in a frost-free freezer) for up to 12 months. Avoid repeated freeze-thaw cycles.
FGF2 (FGFb, fibroblast growth factor basic) belongs to the fibroblast growth factor (FGF) family, and interacts with high-affinity transmembrane receptors to influence cell proliferation and tissue neovascularization. FGF2 exists as five isoforms with distinct intracellular localizations and functions. The 18 kDa isoform is predominantly cytosolic and acts through cell surface receptors, whereas the 22, 22.5, 24 and 34 kDa isoforms are nuclear and may signal independent of transmembrane receptor pathways. In humans, the gene is located on the q arm of chromosome 4. FGF2 has been implicated in diverse biological processes, such as limb and nervous system development, wound healing, and tumor growth. The mRNA for FGF2 contains multiple polyadenylation sites, and is alternatively translated from non-AUG and AUG initiation codons, resulting in five different isoforms with distinct properties. The CUG-initiated isoforms are localized in the nucleus and are responsible for the intracrine effect, whereas, the AUG-initiated form is mostly cytosolic and is responsible for the paracrine and autocrine effects of this FGF. Diseases associated with FGF2 dysfunction include Kaposi Sarcoma and corneal neovascularization.Specifications
| P09038 | |
| 20mM potassium phosphate with 750mM NaCl and no preservative | |
| 19 kDa | |
| Purified | |
| -20°C | |
| <1 EU/μg | |
| FGF2 | |
| 155 aa + 20 aa for N-terminal tag for purification purposes | |
| Protein | |
| Human | |
| Sterile | |
| -20°C | |
| ED50 = 0.6 - 1.1 ng/mL; determined by the dose-dependent proliferation of mouse NIH/3T3 fibroblast cells. | |
| Approved for shipment on Wet or Dry Ice | |
| Growth Factors and Receptors | |
| Neurobiology, Oncology, Stem Cell Research, Microbiology, Differentiation, Angiogenesis |
| Bioactivity | |
| 2247 | |
| Human Heat Stable bFGF | |
| 5 μg | |
| RUO | |
| Basic fibroblast growth factor; basic fibroblast growth factor bFGF; bFGF; FGF; fgf basic; Fgf2; Fgf-2; Fgfb; FGF-b; Fibroblast growth factor; fibroblast growth factor 2; fibroblast growth factor 2 (basic); Fibroblast growth factor basic; HBGF-2; Heparin-binding growth factor 2; H-FGF-b-147; H-FGF-b-154; M-FGF-b; Prostatic growth factor; prostatropin | |
| FGF2 | |
| ED50 = 0.6 - 1.1 ng/mL; determined by the dose-dependent proliferation of mouse NIH/3T3 fibroblast cells. | |
| Unconjugated | |
| Recombinant | |
| 155 aa + 20 aa for N-terminal tag for purification purposes | |
| Carrier-Free | |
| <1 EU/μg | |
| E. coli | |
| Lyophilized | |
| >95% by SDS-PAGE |