missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Gibco™ Human Heat Stable bFGF Recombinant Protein DFS Item

Gibco Heat Stable Recombinant Human Bfgf is a heat stable version of basic FGF (Fibroblast Growth Factor) engineered for greater stability in cell culture conditions. HS bFGF maintains at least 50% more activity after 72 hours, in cell culture conditions than competitor product.

Supplier:  Gibco™ PHG0368

 View more versions of this product

Catalog No. PHG0368


Explore available promotions

Description

Description

Human Heat Stable bFGF Recombinant Protein for Western Blot, IHC, ELISA, Ctrl, FN

  • Superior performance - Retains bioactivity without the use of artificially high concentrations.
  • Easy to use - Direct replacement for your current bFGF allowing for easy substitution in protocols.
  • Advancement without compromise - > 90% sequence homology to native bFGF, leaving heparin and FGF receptor binding sites untouched
Specifications

Specifications

Cell Culture
50 μg
155 aa + 20 aa for N-terminal tag for purification purposes
Heat Stable Recombinant Human bFGF
Sterile
Carrier-Free
Approved for shipment on Wet or Dry Ice
Growth Factors and Receptors
Neurobiology, Oncology, Stem Cell Research, Microbiology, Differentiation, Angiogenesis
19 kDa
Less than 1 EU/ug
ED50 range = 0.6-1.1 ng⁄mL
Human
Store the lyophilized protein at -20°C, preferably desiccated.
< 1 EU/μg
E. coli
Lyophilized
Greater than 95% by SDS-PAGE
Videos
SDS
Documents

Documents

Promotions

Promotions