missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
MP Biomedicals™ Serum, Human, Pooled
Used as a media component to neutralize cell free virus.
Supplier: MP Biomedicals™ 092930149
Description
Applications: Human pooled serum is used as a media component to neutralize cell free virus. It's used as a reagent in liposome binding assay and in preparation of sperm.
Product Description: This serum is obtained from pooled, normal clotted human blood.
Specifications
Golden Brown | |
Liquid | |
100 mL | |
Media component for cell free virus, liposome binding assay, preparation of sperm |
Store at -20 °C. | |
Human Serum | |
Sterile | |
7 to 8 |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.