missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human L-Selectin (CD62L) (Soluble) ELISA Kit

ELISA

$856.00 - $5960.00

Specifications

Accession Number P14151
Assay Range 0.4 to 25.0 ng/mL
Assay Sensitivity 0.198 ng/mL
Conjugate HRP
Product Type ELISA
View More Specs

Includes: Aluminium pouch(es) with a Microwell Plate coated with monoclonal antibody to human sL-selectin
HRP-Conjugate anti-human sL-selectin monoclonal antibody, ready to use
Human sL-selectin Standard lyophilized, 50ng/mL upon reconstitution
Sample Diluent
Wash Buffer Concentrate 20x (PBS with 1% Tween 20)
Substrate Solution (tetramethyl-benzidine)
Stop Solution (1M Phosphoric acid)
Blue-Dye
Green-Dye
Adhesive Films

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5017974
View Documents
Invitrogen™
BMS206TEN
10 x 96 Tests
Each for $5,960.00
Only null left
 
5017967
View Documents
Invitrogen™
BMS206
96 Tests
Each for $856.00
Only null left
 
Description

Description

The Human L-Selectin (CD62L) ELISA quantitates Hu L-Selectin in human serum, plasma or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu L-Selectin. Principle of the method The Human L-Selectin solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.

The human CD62L is a 74-95 kDa glycoprotein member of the selectin family of adhesion receptors. L-Selectin is comprised of an aminoterminal C-type lectin binding domain, an epidermal growth factor-like domain, two short consensus repeat (SCR) sequences homologous to those found in complement binding proteins, a short spacer region, a transmembrane region and a short cytoplasmic region. Human CD62L (L-Selectin) is constitutively expressed on all classes of leukocytes including lymphocytes (except a substantial population of memory T cells), monocytes and polymorphonuclear cells.
Specifications

Specifications

P14151
0.198 ng/mL
ELISA
Human
Colorimetric Microplate Reader
CD62L,LAM1,LECAM1,LEU8,LNHR,LSEL,LYAM1,PLNHR,TQ1
3.7
HRP
Plasma 10 μL, Serum 10 μL, Supernatant 10 μL
Lymphocyte adhesion molecule 1, CD62L, Leu-8
1 hrs 20 mins
0.4 to 25.0 ng/mL
HRP
Plasma, Serum, Supernatant
ELISA
6402
4.2%
Pre-coated 96 well plate, Standard, Sample Diluent, Assay Buffer concentrate, HRP-conjugated Detection Antibody, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers
RUO
2°C to 8°C
L-Selectin (CD62L)
2 hrs 10 mins
Videos
SDS
missing translation for 'documents'

missing translation for 'documents'