missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Corning™ PYREX™ Hybridization Tubes
For use in hybridization incubators with rotators
$426.83 - $472.59
Specifications
Closure Material | Polypropylene |
---|---|
Color | Orange |
Diameter (English) Inner | 1.37 in. |
Diameter (Metric) Inner | 35 mm |
Diameter (Metric) Outer | 35 mm |
Catalog Number | Mfr. No. | Length (Metric) | Price | Quantity | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Length (Metric) | Price | Quantity | ||||||
11-381-16B
|
Corning™
7995150 |
150 mm |
Case of 2 Each for $426.83
|
|
||||||
11-381-16C
|
Corning™
7995300 |
300 mm |
Case of 2 Each for $472.59
|
|
||||||
Description
35 mm I.D. tubes available in three standard lengths for use in hybridization incubators with rotators. A more convenient method requiring smaller probe solution volumes. Comes with Corning™ standard GL 45 orange plug-seal cap incorporating a silicone O-ring to ensure a leak-resistant seal. The optional Corning vented membrane cap is available for low temperature assays.
Cat. No. 1395-OR is replacement part for silicone o-ring.
Cat. No. 1395-45LTC is replacement part for screw cap with plug seal.
Specifications
Polypropylene | |
1.37 in. | |
35 mm | |
Borosilicate Glass | |
10 mL | |
Plug-seal cap, O-ring |
Orange | |
35 mm | |
Plug Seal | |
Hybridization Tube | |
Hybridization incubators with rotators |