MP Biomedicals™ Hybrisol VII
Hybrisol VII
Supplier:  MP Biomedicals™ 11RIST139010
Catalog No. MPRIST13901
Explore available promotions
Description
Description
  • Hybrisol VII for DNA FISH.
  • It facilitates probe hybridization to DNA in whole cells, metaphase chromosomes, or interphase nuclei.
Specifications
Specifications
Hybridization Solution
10 x 1 mL