Cytiva HyClone™ Cryopreservation Media
Animal origin-free cryopreservation media for general use
Supplier:  Cytiva SR3000102
Catalog No. SR3000102
Explore available promotions
Description
Description
  • Serum-free, animal origin-free for cryogenic storage for standard cell lines
  • Meets industry standard for viable cell recovery per vial; outperforms other cryo media solution in the market for viable cell recovery per vial
  • Provided as a 2X solution; 0.2μm sterile filtered
  • For further manufacturing or research use. Not for diagnostic or therapeutic use.
Specifications
Specifications
100 mL