missing translation for 'onlineSavingsMsg'
Learn More

Cytiva HyClone™ Characterized Fetal Bovine Serum (U.S.)

Used as serum-supplement for the in vitro cell culture of eukaryotic cells

Supplier:  Cytiva SH3007103

 View more versions of this product

Catalog No. SH3007103


Only null left
Explore available promotions

Description

Description

  • Enhance growth of cells
  • Tested for endotoxin, hemoglobin, and additional biochemical assays
  • Filtered through serial 100 nm pore-size rated filters
Specifications

Specifications

Albumin, Alkaline Phosphatase, Lactate Dehydrogenase, Osmolality, Chloride, pH, Potassium, Glucose, SGPT, Sodium, TIBC, Alkaline Phosphatase, Total Bilirubin, Total Protein, Blood Urea Nitrogen, Calcium, Chloride, Creatinine, Gamma Globulin, Glucose, IgG, Inorganic Phosphorus
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2 to 8°C.
500 mL
Sequential Triple-filtered, 100 nm
None
≤ 25 EU/mL
≤ 25 mg/dL
No
Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities
Fetal Bovine Serum
Sterile
Characterized
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus
No
HyClone
USDA Inspected
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions