missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Cytiva Hyclone™ Defined Fetal Bovine Serum (U.S.)
Used as serum-supplement for the in vitro cell culture of eukaryotic cells
Supplier: Cytiva SH3007002HI
Description
- Maximize cell culture growth
- Tested for endotoxin, hemoglobin, and additional biochemical assays
- Filtered through serial 40 nm pore-size rated filters
Specifications
Progesterone,Insulin,Cholesterol,HDL,LDL,Triglycerides,Gamma Globulin,Phospholipids,Alkaline Phosphatase,Lactate Dehydrogenase,SGPT,SGOT,B1,B2,B6,Niacin,Biotin,Retinol,Carotene,Tocopherol,B12,Folate,Vitamin C,1,25-dihydroxy vitamin D,pH,Total Protein,Albumin,Blood Urea Nitrogen,Creatinine,Total Bilirubin,Inorganic Phosphorus,Selenium,Sodium,Potassium,Calcium,Chloride,Magnesium,Glucose,Copper,Osmolality,Iron,TIBC,Percent Saturation,IgG | |
Sera should be stored at -10°C or lower. Once thawed, sera should be stored at 2 to 8°C. | |
Fetal Bovine Serum | |
Sterile | |
Defined | |
Bluetongue, Bovine Adenovirus, Bovine Parvovirus, Bovine RSV, BVDV, Cytopathogenic Agents, Hemadsorbing Agents, Rabies Virus, Reovirus | |
Yes | |
HyClone™ | |
USDA Inspected |
Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities | |
United States | |
100 mL | |
Sequential-filtered, 40 nm | |
Heat Inactivated | |
≤ 10 EU/mL | |
≤ 10 mg/dL | |
No |